Assay Details
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244327.1 | NP_001231256.1 | 1 - 2 | 124 | 119 | |
XM_005668398.3 | XP_005668455.1 | 217 | 119 | |||
XM_013980494.2 | XP_013835948.1 | 230 | 119 | |||
XM_013980495.2 | XP_013835949.1 | 212 | 119 | |||
XM_013980496.2 | XP_013835950.1 | 235 | 119 | |||
GenBank mRNA | AK231303.1 | - | 1 - 2 | 117 | 119 | |
AK232427.1 | - | 1 - 2 | 146 | 119 | ||
AK235737.1 | - | 1 - 2 | 123 | 119 | ||
AK239065.1 | - | 1 - 2 | 117 | 119 | ||
AK348844.1 | - | 1 - 2 | 117 | 119 | ||
AK352029.1 | - | 1 - 2 | 117 | 119 | ||
AK395116.1 | - | 1 - 2 | 122 | 119 | ||
BW982187.1 | - | 1 - 2 | 117 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
