Assay Details
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244327.1 | NP_001231256.1 | 2 - 3 | 244 | 67 | |
XM_005668398.3 | XP_005668455.1 | 337 | 67 | |||
XM_013980494.2 | XP_013835948.1 | 350 | 67 | |||
XM_013980495.2 | XP_013835949.1 | 332 | 67 | |||
XM_013980496.2 | XP_013835950.1 | 355 | 67 | |||
GenBank mRNA | AK231303.1 | - | 2 - 3 | 237 | 67 | |
AK232427.1 | - | 2 - 3 | 266 | 67 | ||
AK235737.1 | - | 2 - 3 | 243 | 67 | ||
AK236832.1 | - | 2 - 3 | 237 | 67 | ||
AK239065.1 | - | 2 - 3 | 237 | 67 | ||
AK348844.1 | - | 2 - 3 | 237 | 67 | ||
AK352029.1 | - | 2 - 3 | 237 | 67 | ||
AK395116.1 | - | 2 - 3 | 242 | 67 | ||
AK395223.1 | - | 2 - 3 | 243 | 67 | ||
BW982187.1 | - | 2 - 3 | 237 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
