Assay Details
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244327.1 | NP_001231256.1 | 4 - 5 | 486 | 82 | |
XM_005668398.3 | XP_005668455.1 | 579 | 82 | |||
XM_013980494.2 | XP_013835948.1 | 592 | 82 | |||
XM_013980495.2 | XP_013835949.1 | 574 | 82 | |||
XM_013980496.2 | XP_013835950.1 | 597 | 82 | |||
GenBank mRNA | AK232427.1 | - | 4 - 5 | 508 | 82 | |
AK235737.1 | - | 4 - 5 | 485 | 82 | ||
AK236832.1 | - | 4 - 5 | 479 | 82 | ||
AK239065.1 | - | 4 - 5 | 479 | 82 | ||
AK352029.1 | - | 4 - 5 | 479 | 82 | ||
AK395116.1 | - | 4 - 5 | 484 | 82 | ||
AK395223.1 | - | 4 - 5 | 485 | 82 | ||
AK397948.1 | - | 2 - 3 | 180 | 82 | ||
AK399378.1 | - | 1 - 2 | 123 | 82 | ||
BW982187.1 | - | 4 - 5 | 479 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
