Assay Details
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244327.1 | NP_001231256.1 | 5 - 6 | 536 | 81 | |
XM_005668398.3 | XP_005668455.1 | 629 | 81 | |||
XM_013980494.2 | XP_013835948.1 | 642 | 81 | |||
XM_013980495.2 | XP_013835949.1 | 624 | 81 | |||
XM_013980496.2 | XP_013835950.1 | 647 | 81 | |||
GenBank mRNA | AK231303.1 | - | 6 - 7 | 654 | 81 | |
AK235737.1 | - | 5 - 6 | 535 | 81 | ||
AK236832.1 | - | 5 - 6 | 529 | 81 | ||
AK239065.1 | - | 5 - 6 | 529 | 81 | ||
AK352029.1 | - | 5 - 6 | 529 | 81 | ||
AK395116.1 | - | 5 - 6 | 534 | 81 | ||
AK399378.1 | - | 2 - 3 | 173 | 81 | ||
BW982187.1 | - | 5 - 6 | 529 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
