Assay Details
| Gene Symbol: CRLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244329.1 | NP_001231258.1 | 5 - 6 | 897 | 75 | |
| XM_005669062.3 | XP_005669119.2 | 922 | 75 | |||
| XM_021066135.1 | XP_020921794.1 | 925 | 75 | |||
| GenBank mRNA | CJ019492.1 | - | 2 - 3 | 345 | 75 | |
More Information
Set Membership: |
|

CRLF3
Genomic Map