Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244333.1 | NP_001231262.1 | 6 - 7 | 750 | 104 | |
| XM_021079875.1 | XP_020935534.1 | 770 | 104 | |||
| XM_021079878.1 | XP_020935537.1 | 2068 | 104 | |||
| XM_021079882.1 | XP_020935541.1 | 1613 | 104 | |||
| XM_021079888.1 | XP_020935547.1 | 852 | 104 | |||
| GenBank mRNA | AK232442.1 | - | 6 - 7 | 753 | 104 | |
| AK233387.1 | - | 6 - 7 | 736 | 104 | ||
| AK240422.1 | - | 6 - 7 | 750 | 104 | ||
| AK396496.1 | - | 6 - 7 | 1041 | 104 | ||
| AK397779.1 | - | 6 - 7 | 762 | 104 | ||
| AK398071.1 | - | 6 - 7 | 702 | 104 | ||
| AK398953.1 | - | 7 - 8 | 833 | 104 | ||
More Information
Set Membership: |
|

GRAMD2B
Genomic Map