Assay Details
| Gene Symbol: SLC35D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244362.1 | NP_001231291.1 | 5 - 6 | 469 | 79 | |
| XM_021063996.1 | XP_020919655.1 | 659 | 79 | |||
| XM_021063997.1 | XP_020919656.1 | 587 | 79 | |||
| GenBank mRNA | AK238334.1 | - | 5 - 6 | 469 | 79 | |
More Information
Set Membership: |
|

SLC35D2
Genomic Map