Assay Details
| Gene Symbol: CDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244385.1 | NP_001231314.1 | 1 - 2 | 208 | 91 | |
| GenBank mRNA | AK346675.1 | - | 1 - 2 | 186 | 91 | |
| AY610472.1 | - | 1 - 2 | 208 | 91 | ||
More Information
Set Membership: |
|

CDA
Genomic Map