Assay Details
| Gene Symbol: PPID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244386.1 | NP_001231315.1 | 4 - 5 | 600 | 65 | |
| XM_021100359.1 | XP_020956018.1 | 1199 | 65 | |||
| GenBank mRNA | AK344118.1 | - | 4 - 5 | 595 | 65 | |
| AY609657.1 | - | 4 - 5 | 600 | 65 | ||
More Information
Set Membership: |
|

PPID
Genomic Map