Assay Details
| Gene Symbol: PDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244398.1 | NP_001231327.1 | 2 - 3 | 162 | 87 | |
| GenBank mRNA | AK236324.1 | - | 2 - 3 | 162 | 87 | |
| AK393624.1 | - | 2 - 3 | 195 | 87 | ||
More Information
Set Membership: |
|

PDHB
Genomic Map