Assay Details
Gene Symbol: PPWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244421.1 | NP_001231350.1 | 1 - 2 | 216 | 74 | |
XM_005672480.3 | XP_005672537.1 | 233 | 74 | |||
XM_021076546.1 | XP_020932205.1 | 233 | 74 | |||
XM_021076547.1 | XP_020932206.1 | 222 | 74 | |||
GenBank mRNA | AK344693.1 | - | 1 - 2 | 216 | 74 | |
FS698804.1 | - | 1 - 2 | 201 | 74 | ||
JN007383.1 | - | 1 - 2 | 202 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
