Assay Details
Gene Symbol: GAL3ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244429.1 | NP_001231358.1 | 1 - 2 | 295 | 83 | |
XM_005670826.3 | XP_005670883.1 | 3040 | 83 | |||
GenBank mRNA | AK231249.1 | - | 1 - 2 | 295 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244429.1, XM_005670826.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
