Assay Details
| Gene Symbol: MRPL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244447.1 | NP_001231376.1 | 5 - 6 | 752 | 68 | |
| NM_001244448.1 | NP_001231377.1 | 5 - 6 | 629 | 68 | ||
| XM_005663256.3 | XP_005663313.1 | 740 | 68 | |||
| XM_005663257.2 | XP_005663314.1 | 799 | 68 | |||
| GenBank mRNA | AK231970.1 | - | 5 - 6 | 752 | 68 | |
| AK233885.1 | - | 5 - 6 | 698 | 68 | ||
| AK346764.1 | - | 5 - 6 | 707 | 68 | ||
| AK394983.1 | - | 5 - 6 | 621 | 68 | ||
| AY609955.1 | - | 5 - 6 | 694 | 68 | ||
More Information
Set Membership: |
|

MRPL24
Genomic Map