Assay Details
| Gene Symbol: PRDX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244474.1 | NP_001231403.1 | 4 - 5 | 494 | 85 | |
| GenBank mRNA | AJ963214.1 | - | 1 - 2 | 38 | 85 | |
| AK349436.1 | - | 4 - 5 | 494 | 85 | ||
| AK390553.1 | - | 4 - 5 | 443 | 85 | ||
| AK392270.1 | - | 4 - 5 | 462 | 85 | ||
| AK400020.1 | - | 4 - 5 | 494 | 85 | ||
| AY610343.1 | - | 4 - 5 | 430 | 85 | ||
| GQ913659.1 | - | 4 - 5 | 448 | 85 | ||
More Information
Set Membership: |
|

PRDX2
Genomic Map