Assay Details
Gene Symbol: WASHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244483.1 | NP_001231412.1 | 2 - 3 | 182 | 104 | |
GenBank mRNA | AK236494.1 | - | 2 - 3 | 182 | 104 | |
AK399336.1 | - | 2 - 3 | 202 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
