Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244483.1 | NP_001231412.1 | 4 - 5 | 445 | 91 | |
| GenBank mRNA | AK236494.1 | - | 4 - 5 | 445 | 91 | |
| AK399336.1 | - | 4 - 5 | 465 | 91 | ||
More Information
Set Membership: |
|

WASHC1
Genomic Map