Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244483.1 | NP_001231412.1 | 6 - 7 | 715 | 69 | |
| GenBank mRNA | AK236494.1 | - | 6 - 7 | 715 | 69 | |
| AK399336.1 | - | 6 - 7 | 735 | 69 | ||
More Information
Set Membership: |
|

WASHC1
Genomic Map