Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244483.1 | NP_001231412.1 | 10 - 11 | 1306 | 63 | |
| GenBank mRNA | AK236494.1 | - | 10 - 11 | 1306 | 63 | |
| AK399336.1 | - | 10 - 11 | 1202 | 63 | ||
| CN158973.1 | - | 4 - 5 | 434 | 63 | ||
| EW670076.2 | - | 1 - 2 | 89 | 63 | ||
More Information
Set Membership: |
|

WASHC1
Genomic Map