Assay Details
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244527.1 | NP_001231456.1 | 3 - 4 | 640 | 108 | |
GenBank mRNA | AY610280.1 | - | 3 - 4 | 474 | 108 | |
EW454624.2 | - | 3 - 4 | 481 | 108 | ||
FD642407.1 | - | 3 - 4 | 640 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
