Assay Details
| Gene Symbol: PRDX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244531.1 | NP_001231460.1 | 2 - 3 | 196 | 97 | |
| GenBank mRNA | AK235228.1 | - | 2 - 3 | 238 | 97 | |
| AK235800.1 | - | 2 - 3 | 237 | 97 | ||
| AK348125.1 | - | 2 - 3 | 198 | 97 | ||
| AK351883.1 | - | 2 - 3 | 196 | 97 | ||
| AK392207.1 | - | 2 - 3 | 226 | 97 | ||
| AK392777.1 | - | 2 - 3 | 206 | 97 | ||
More Information
Set Membership: |
|

PRDX3
Genomic Map