Assay Details
| Gene Symbol: CDKN2AIPNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244537.1 | NP_001231466.1 | 2 - 3 | 345 | 100 | |
| GenBank mRNA | AK343995.1 | - | 2 - 3 | 191 | 100 | |
| CK466548.1 | - | 2 - 3 | 288 | 100 | ||
More Information
Set Membership: |
|

CDKN2AIPNL
Genomic Map