Assay Details
Gene Symbol: ARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244544.1 | NP_001231473.1 | 1 - 2 | 198 | 121 | |
GenBank mRNA | AK346576.1 | - | 1 - 2 | 198 | 121 | |
AK348002.1 | - | 1 - 2 | 198 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
