Assay Details
Gene Symbol: ARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244544.1 | NP_001231473.1 | 3 - 4 | 402 | 69 | |
XM_021098268.1 | XP_020953927.1 | 392 | 69 | |||
XM_021098270.1 | XP_020953929.1 | 532 | 69 | |||
XM_021098272.1 | XP_020953931.1 | 456 | 69 | |||
GenBank mRNA | AK348002.1 | - | 3 - 4 | 402 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021098268.1, XM_021098270.1, XM_021098272.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
