Assay Details
| Gene Symbol: AIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244556.1 | NP_001231485.1 | 2 - 3 | 485 | 66 | |
| NM_001244557.1 | NP_001231486.1 | 2 - 3 | 461 | 66 | ||
| GenBank mRNA | AK234127.1 | - | 2 - 3 | 66 | ||
| AK234283.1 | - | 2 - 3 | 66 | |||
| AK238255.1 | - | 2 - 3 | 66 | |||
| AK347601.1 | - | 2 - 3 | 66 | |||
| AK398817.1 | - | 2 - 3 | 66 | |||
| BW976518.1 | - | 2 - 3 | 66 | |||
| CJ008617.1 | - | 2 - 3 | 472 | 66 | ||
| CK454751.1 | - | 1 - 2 | 87 | 66 | ||
More Information
Set Membership: |
|

AIMP2
Genomic Map