Assay Details
| Gene Symbol: AIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244556.1 | NP_001231485.1 | 3 - 4 | 707 | 64 | |
| NM_001244557.1 | NP_001231486.1 | 3 - 4 | 683 | 64 | ||
| GenBank mRNA | AK234127.1 | - | 3 - 4 | 654 | 64 | |
| AK234283.1 | - | 3 - 4 | 691 | 64 | ||
| AK238255.1 | - | 3 - 4 | 682 | 64 | ||
| AK347601.1 | - | 3 - 4 | 678 | 64 | ||
| AK398817.1 | - | 3 - 4 | 707 | 64 | ||
| BW976518.1 | - | 3 - 4 | 707 | 64 | ||
| CJ008617.1 | - | 3 - 4 | 694 | 64 | ||
| CK454751.1 | - | 2 - 3 | 309 | 64 | ||
More Information
Set Membership: |
|

AIMP2
Genomic Map