Assay Details
Gene Symbol: NAGPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244574.1 | NP_001231503.1 | 3 - 4 | 704 | 72 | |
XM_005662151.3 | XP_005662208.1 | 734 | 72 | |||
XM_005662153.3 | XP_005662210.1 | 734 | 72 | |||
XM_013995664.2 | XP_013851118.1 | 734 | 72 | |||
XM_013995665.2 | XP_013851119.1 | 734 | 72 | |||
XM_021085657.1 | XP_020941316.1 | 734 | 72 | |||
XM_021085658.1 | XP_020941317.1 | 734 | 72 | |||
GenBank mRNA | AK237573.1 | - | 3 - 4 | 687 | 72 | |
AK389600.1 | - | 1 - 2 | 142 | 72 | ||
AK393958.1 | - | 3 - 4 | 694 | 72 | ||
DB789560.1 | - | 3 - 4 | 687 | 72 | ||
FD628352.1 | - | 3 - 4 | 704 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
