Assay Details
| Gene Symbol: EIF3I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244624.1 | NP_001231553.1 | 9 - 10 | 860 | 62 | |
| GenBank mRNA | AK232367.1 | - | 9 - 10 | 860 | 62 | |
| AK394444.1 | - | 9 - 10 | 863 | 62 | ||
| AK397989.1 | - | 9 - 10 | 850 | 62 | ||
More Information
Set Membership: |
|

EIF3I
Genomic Map