Assay Details
| Gene Symbol: NDUFA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244646.1 | NP_001231575.1 | 3 - 4 | 346 | 68 | |
| GenBank mRNA | AK400086.1 | - | 3 - 4 | 276 | 68 | |
| CK466967.1 | - | 3 - 4 | 232 | 68 | ||
| EW150993.2 | - | 3 - 4 | 350 | 68 | ||
More Information
Set Membership: |
|

NDUFA13
Genomic Map