Assay Details
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244694.1 | NP_001231623.1 | 3 - 4 | 318 | 93 | |
XM_005653456.2 | XP_005653513.1 | 294 | 93 | |||
GenBank mRNA | AK399671.1 | - | 3 - 4 | 313 | 93 | |
EW455534.2 | - | 3 - 4 | 267 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
