Assay Details
| Gene Symbol: PVRIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244699.1 | NP_001231628.1 | 3 - 4 | 1280 | 68 | |
| XM_005661897.3 | XP_005661954.1 | 927 | 68 | |||
| XM_021085659.1 | XP_020941318.1 | 945 | 68 | |||
| GenBank mRNA | AK233870.1 | - | 2 - 3 | 697 | 68 | |
| AK240369.1 | - | 3 - 4 | 1280 | 68 | ||
More Information
Set Membership: |
|

PVRIG
Genomic Map