Assay Details
| Gene Symbol: MOCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244725.1 | NP_001231654.1 | 5 - 6 | 589 | 92 | |
| XM_021076548.1 | XP_020932207.1 | 515 | 92 | |||
| GenBank mRNA | AK344333.1 | - | 5 - 6 | 589 | 92 | |
| AY609538.1 | - | 5 - 6 | 568 | 92 | ||
More Information
Set Membership: |
|

MOCS2
Genomic Map