Assay Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244728.1 | NP_001231657.1 | 1 - 2 | 220 | 61 | |
| GenBank mRNA | AK349988.1 | - | 1 - 2 | 220 | 61 | |
More Information
Set Membership: |
|

VMO1
Genomic Map