Assay Details
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244742.1 | NP_001231671.1 | 7 - 8 | 698 | 79 | |
GenBank mRNA | AY609561.1 | - | 7 - 8 | 701 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
