Assay Details
| Gene Symbol: MMACHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244776.1 | NP_001231705.1 | 1 - 2 | 99 | 63 | |
| GenBank mRNA | AK395110.1 | - | 1 - 2 | 98 | 63 | |
| BP145298.1 | - | 1 - 2 | 99 | 63 | ||
More Information
Set Membership: |
|

MMACHC
Genomic Map