Assay Details
| Gene Symbol: USB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244803.1 | NP_001231732.1 | 3 - 4 | 532 | 64 | |
| GenBank mRNA | AK237059.1 | - | 3 - 4 | 532 | 64 | |
More Information
Set Membership: |
|

USB1
Genomic Map