Assay Details
| Gene Symbol: NAA50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244807.1 | NP_001231736.1 | 4 - 5 | 627 | 122 | |
| XM_005670201.3 | XP_005670258.1 | 656 | 122 | |||
| GenBank mRNA | AK234091.1 | - | 4 - 5 | 627 | 122 | |
| AK236454.1 | - | 4 - 5 | 638 | 122 | ||
| AY609740.2 | - | 4 - 5 | 580 | 122 | ||
More Information
Set Membership: |
|

NAA50
Genomic Map