Assay Details
| Gene Symbol: MRPS34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244832.1 | NP_001231761.1 | 2 - 3 | 380 | 86 | |
| GenBank mRNA | AK345219.1 | - | 2 - 3 | 380 | 86 | |
| AK394377.1 | - | 2 - 3 | 387 | 86 | ||
More Information
Set Membership: |
|

MRPS34
Genomic Map