Assay Details
| Gene Symbol: PRRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244836.1 | NP_001231765.1 | 5 - 6 | 858 | 82 | |
| XM_013994445.2 | XP_013849899.1 | 849 | 82 | |||
| GenBank mRNA | AK349291.1 | - | 5 - 6 | 858 | 82 | |
| AK390141.1 | - | 5 - 6 | 914 | 82 | ||
More Information
Set Membership: |
|

PRRG4
Genomic Map