Assay Details
| Gene Symbol: TAF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243639.1 | NP_001230568.1 | 1 - 2 | 76 | 108 | |
| GenBank mRNA | FS644746.1 | - | 1 - 2 | 76 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243639.1 transcripts.
Set Membership: |
|

TAF13
Genomic Map