Assay Details
| Gene Symbol: NDUFA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185178.1 | NP_001172107.1 | 3 | 620 | 97 | |
| GenBank mRNA | AK346757.1 | - | 3 | 617 | 97 | |
| CJ011531.1 | - | 3 | 618 | 97 | ||
More Information
Set Membership: |
|

NDUFA6
Genomic Map