Assay Details
| Gene Symbol: FKBP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243807.1 | NP_001230736.1 | 5 | 820 | 128 | |
| GenBank mRNA | AK234292.1 | - | 5 | 856 | 128 | |
| AK347660.1 | - | 5 | 826 | 128 | ||
More Information
Set Membership: |
|

FKBP10
Genomic Map