Assay Details
| Gene Symbol: SUOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244498.1 | NP_001231427.1 | 4 | 1924 | 129 | |
| XM_021091097.1 | XP_020946756.1 | 2083 | 129 | |||
| XM_021091098.1 | XP_020946757.1 | 2022 | 129 | |||
| GenBank mRNA | AK233618.1 | - | 4 | 1951 | 129 | |
| AK394822.1 | - | 4 | 1951 | 129 | ||
| AK397998.1 | - | 4 | 2185 | 129 | ||
More Information
Set Membership: |
|

SUOX
Genomic Map