Assay Details
| Gene Symbol: CTSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089893.1 | XP_020945552.1 | 3603 | 81 | ||
| GenBank mRNA | AK230540.1 | - | 7 - 8 | 997 | 81 | |
| AK239742.1 | - | 7 - 8 | 978 | 81 | ||
| AK389756.1 | - | 7 - 8 | 1039 | 81 | ||
| AK389844.1 | - | 7 - 8 | 1039 | 81 | ||
| AK391540.1 | - | 7 - 8 | 970 | 81 | ||
More Information
Set Membership: |
|

CTSS
Genomic Map