Assay Details
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005671638.3 | XP_005671695.2 | 348 | 68 | ||
XM_021076421.1 | XP_020932080.1 | 316 | 68 | |||
GenBank mRNA | AK240430.1 | - | 2 - 3 | 298 | 68 | |
AK351903.1 | - | 2 - 3 | 298 | 68 | ||
AK394341.1 | - | 2 - 3 | 331 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
