Assay Details
Gene Symbol: UFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125654.4 | XP_003125702.1 | 811 | 64 | ||
XM_003125655.4 | XP_003125703.1 | 322 | 64 | |||
XM_021089535.1 | XP_020945194.1 | 194 | 64 | |||
GenBank mRNA | AK239911.1 | - | 3 - 4 | 375 | 64 | |
AK396135.1 | - | 3 - 4 | 377 | 64 | ||
AY610495.1 | - | 3 - 4 | 283 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
