Assay Details
Gene Symbol: MIR505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038578.1 | 1 | 40 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATPase phospholipid transporting 11C | ATP11C | Sense | Chr. X - 114394436 - 114575658 on NCBI Build Sscrofa11.1 | ||
Downstream | protein transport protein SEC31-like | LOC110257749 | 12.0kb | Anti-sense | Chr. X - 114575980 - 115026005 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
