Assay Details
| Gene Symbol: MIR19B-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035400.1 | 1 | 40 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-20a | MIR20A |
MIR20, MIRN20, MIRN20A, ssc-mir-20, ssc-mir-20a |
-59.0 | Sense | Chr. 11 - 60735109 - 60735179 on NCBI Build Sscrofa11.1 |
| Downstream | microRNA mir-92a-1 | MIR92A-1 | ssc-mir-92a-1 | 41.0 | Sense | Chr. 11 - 60735358 - 60735437 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR19B-1
Genomic Map