Assay Details
| Gene Symbol: MIR18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031541.2 | 1 | 53 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-17 | MIR17 | ssc-mir-17 | -59.0 | Sense | Chr. 11 - 60734656 - 60734732 on NCBI Build Sscrofa11.1 | 
| Downstream | microRNA mir-19a | MIR19A | MIRN19A, ssc-mir-19a | 57.0 | Sense | Chr. 11 - 60734939 - 60735020 on NCBI Build Sscrofa11.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Detect Genomic DNA  Sus scrofa  Both primers and probe map within a single exon | 
 Back To Top
 Back To Top
 MIR18A
MIR18A Genomic Map
Genomic Map Back To Top
 Back To Top