Assay Details
Gene Symbol: LOC100152407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033705.1 | 2 - 3 | 425 | 75 | ||
GenBank mRNA | AK231161.1 | - | 2 - 3 | 424 | 75 | |
AK351713.1 | - | 2 - 3 | 425 | 75 | ||
AK394859.1 | - | 2 - 3 | 422 | 75 | ||
AK400180.1 | - | 2 - 3 | 422 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromodomain helicase DNA binding protein 2 | CHD2 | -13.2kb | Sense | Chr. 7 - 85980759 - 86107886 on NCBI Build Sscrofa11.1 | |
Downstream | family with sequence similarity 174 member B | FAM174B | 60.2kb | Anti-sense | Chr. 7 - 86181970 - 86356953 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
