Assay Details
| Gene Symbol: MIR30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035368.1 | 1 | 40 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-30d | MIR30D | ssc-mir-30d | -4.0kb | Sense | Chr. 4 - 6948669 - 6948747 on NCBI Build Sscrofa11.1 |
| Downstream | zinc finger and AT-hook domain containing | ZFAT | 16.8kb | Sense | Chr. 4 - 6969692 - 7174989 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR30B
Genomic Map